Skip to Content
Merck
All Photos(2)

Key Documents

330957

Sigma-Aldrich

2-(Dimethylamino)ethyl acrylate

contains <2,000 ppm MEHQ as inhibitor, 98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$167.00
500 ML
$427.00

$167.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$167.00
500 ML
$427.00

About This Item

Linear Formula:
H2C=CHCO2CH2CH2N(CH3)2
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$167.00


Available to ship on22 April 2025Details


Request a Bulk Order

vapor pressure

0.35 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

contains

<2,000 ppm MEHQ as inhibitor

refractive index

n20/D 1.438 (lit.)

bp

64 °C/12 mmHg (lit.)

density

0.943 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)CCOC(=O)C=C

InChI

1S/C7H13NO2/c1-4-7(9)10-6-5-8(2)3/h4H,1,5-6H2,2-3H3

InChI key

DPBJAVGHACCNRL-UHFFFAOYSA-N

Application

2-(Dimethylamino)ethyl acrylate (AmAc) are aminoacrylates. AmAc was used to fabricate gold/acrylic polymer nanocomposites.1 2-(dimethylamino)ethyl acrylate (DAEA) can undergo copolymerization with 2-acrylamido-2-methylpropanesulphonic acid (AMPS). [1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of copolymers of 2-(dimethylamino)ethyl acrylate with 2-acrylamido-2-methylpropanesulphonic acid
Aggour YA
Polymer Degradation and Stability, 45(3), 273-276 (1994)
Despoina Giaouzi et al.
Polymers, 12(6) (2020-06-25)
In this work, the synthesis, selective chemical modifications, and self-assembly behavior in aqueous media of a novel poly(2-(dimethylamino)ethyl acrylate)20-b-poly(N-isopropylacrylamide)11-b-poly(oligo ethylene glycol methyl ether acrylate)18 (PDMAEA20-b-PNIPAM11-b-POEGA18) dual-responsive (pH and temperature) and triply hydrophilic amino-based triblock terpolymer are reported. The amine functional
Ali Dinari et al.
Scientific reports, 8(1), 8112-8112 (2018-05-29)
Engineering molecules at nano-scale is a promising approach in targeting and curing diseases. In this research, fabricated new hybrid system called nano-polyplex represents an example of the molecular engineering at nano-scale. Polymer of PDMAEAs with four different molecular weights were
Steven Martens et al.
Nature communications, 9(1), 4451-4451 (2018-10-28)
Sequence-defined macromolecules consist of a defined chain length (single mass), end-groups, composition and topology and prove promising in application fields such as anti-counterfeiting, biological mimicking and data storage. Here we show the potential use of multifunctional sequence-defined macromolecules as a

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. I wanted to know is this a polymer or monomerba

    1 answer
    1. 2-(Dimethylamino)ethyl acrylate is a monomer and contains MEHQ as a polymerization inhibitor.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service