Skip to Content
Merck
All Photos(1)

Key Documents

329916

Sigma-Aldrich

Potassium dideuterium phosphate

98 atom % D

Synonym(s):

Potassium dihydrogen-d2 phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$88.00

$88.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$88.00

About This Item

Linear Formula:
KD2PO4
CAS Number:
Molecular Weight:
138.10
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$88.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

form

solid

technique(s)

NMR: suitable

mp

253 °C (lit.)

mass shift

M+2

SMILES string

[K+].[2H]OP([O-])(=O)O[2H]

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1/i/hD2

InChI key

GNSKLFRGEWLPPA-ZSJDYOACSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yaqi Fu et al.
Biotechnology progress, 37(1), e3075-e3075 (2020-09-02)
Dehaloperoxidase (DHP) catalyzes detoxifying halophenols. It is a heme-containing enzyme using H2 O2 as the oxidant. Heme bleaching from the active site is of great concern. In addition, the interference of DHP by H2 O2 leads to the inactivation of
Samriddh Mudgal et al.
Food chemistry, 197(Pt A), 84-91 (2015-12-01)
We illustrate a method that uses bovine serum albumin (BSA) to control the receptor-accessible part of rebaudioside A (Reb A). The critical micelle concentration (CMC) of Reb A was found to be 4.5 mM and 5 mM at pH 3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service