329266
1,4:3,6-Dianhydro-D-mannitol
95%
Synonym(s):
Isomannide
About This Item
Recommended Products
Assay
95%
form
solid
optical activity
[α]25/D +89°, c = 3 in H2O
mp
80-85 °C (lit.)
SMILES string
O[C@@H]1CO[C@@H]2[C@H](O)CO[C@H]12
InChI
1S/C6H10O4/c7-3-1-9-6-4(8)2-10-5(3)6/h3-8H,1-2H2/t3-,4-,5-,6-/m1/s1
InChI key
KLDXJTOLSGUMSJ-KVTDHHQDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service