Skip to Content
Merck
All Photos(1)

Key Documents

328839

Sigma-Aldrich

Diethylmethoxyborane solution

1.0 M in THF

Synonym(s):

Diethylborinic acid methyl ester, Methyl diethylborinate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$146.00

$146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$146.00

About This Item

Linear Formula:
(C2H5)2BOCH3
CAS Number:
Molecular Weight:
99.97
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$146.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.868 g/mL at 25 °C

SMILES string

CCB(CC)OC

InChI

1S/C5H13BO/c1-4-6(5-2)7-3/h4-5H2,1-3H3

InChI key

FESAXEDIWWXCNG-UHFFFAOYSA-N

Application

Reactant involved in:
  • Studying a catalyst for ring-opening metathesis polymerization / vinyl insertion polymerization
  • Reformatsky / quaternary Claisen condensations
  • Enantioselective synthesis of carba-furanose sugars
  • Borane-mediated control radical polymerization for synthesis of fluoropolymers
  • Homologation reactions with sulfonium ylides
  • Trialkylborane promoted adhesion to low surface energy plastics

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service