Skip to Content
Merck
All Photos(1)

Documents

328316

Sigma-Aldrich

Cyclopentanecarbonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9COCl
CAS Number:
Molecular Weight:
132.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4622 (lit.)

bp

161-162 °C (lit.)

density

1.091 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)C1CCCC1

InChI

1S/C6H9ClO/c7-6(8)5-3-1-2-4-5/h5H,1-4H2

InChI key

WEPUZBYKXNKSDH-UHFFFAOYSA-N

Application

Cyclopentanecarbonyl chloride was used in preparation of:
  • 3′-N-acyl-7-trichloroacetyl-paclitaxels
  • cyclopentanecarboxylic acid 2-[5-(1-cyclopentane carbonyloxy-2-methyl-allyl)-2-oxo-dihydro-furan-3-yl-idene]-ethyl ester
  • cyclopentanecarboxylic acid 2-[5-(1-hydroxy-2-methyl allyl)-2-oxo-dihydro-furan-3-ylidene]-ethyl ester

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Joo Roh et al.
Bioorganic & medicinal chemistry, 10(10), 3145-3151 (2002-08-02)
A series of 3'-N-acyl-paclitaxel analogues 1a-v were synthesized and their cytotoxicities in vitro against several human tumor cell lines examined. It has been shown that distinct correlation between activity and N-acyl-substituent. The appropriate size of N-acyl group was indispensable for
Ester derivatives from tannase-treated prunioside A and their anti-inflammatory activities.
Jun C, et al.
Bull. Korean Chem. Soc., 28(1), 73-73 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service