Skip to Content
Merck
All Photos(1)

Key Documents

328154

Sigma-Aldrich

2,4,5-Trifluorobenzoic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$63.20

$63.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$63.20

About This Item

Linear Formula:
F3C6H2CO2H
CAS Number:
Molecular Weight:
176.09
Beilstein:
3257609
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$63.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

94-96 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1cc(F)c(F)cc1F

InChI

1S/C7H3F3O2/c8-4-2-6(10)5(9)1-3(4)7(11)12/h1-2H,(H,11,12)

InChI key

AKAMNXFLKYKFOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,5-Trifluorobenzoic acid is an important quinolone antibacterial intermediate.[1]

Application

2,4,5-Trifluorobenzoic acid was used in preparation of 5-fluoro-2-methoxy-4-methylaminbenzamide.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yoshimi Hirokawa et al.
Chemical & pharmaceutical bulletin, 50(7), 941-959 (2002-07-20)
In search of a dopamine D2 and serotonin 5-HT3 receptors dual antagonist as a potential broad antiemetic agent, a number of benzamides were prepared from 4-amino-5-chloro-2-methoxybenzoic acid derivatives and 6-amino-1,4-dialkylhexahydro-1,4-diazepines and evaluated for their binding affinity for the dopamine D2
An Expedient Route to the Quinolone Antibacterial Intermediate, 2, 4, 5-Trifluorobenzoic Acid.
O'Reilly NJ, et al.
Synlett, 1990(10), 609-610 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service