Skip to Content
Merck
All Photos(2)

Key Documents

324906

Sigma-Aldrich

4,5,6,7-Tetrahydroindole

98%

Synonym(s):

2,3-Tetramethylenepyrrole, Cyclohex[b]pyrrole, NSC 122455

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$478.00

$478.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$478.00

About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
Beilstein:
108853
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$478.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

53-57 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CCc2[nH]ccc2C1

InChI

1S/C8H11N/c1-2-4-8-7(3-1)5-6-9-8/h5-6,9H,1-4H2

InChI key

KQBVVLOYXDVATK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,5,6,7-Tetrahydroindoles, due to their easy aromatization, are good intermediates to synthesize indoles.[1] 4,5,6,7-Tetrahydroindole on condensation with cyanoacetate leads to 1-ethylthio-2-cyano-4,5,6,7-tetrahydrocyclohexa-[c]-3H-pyrrolizin-3-one.[2]

Application

4,5,6,7-Tetrahydroindole was used as reactant in:
  • synthesis of ethyl 3-(4,5,6,7-tetrahydroindol-2-yl)-2-propynoate[1]
  • preparation of BODIPY dyes[3]
  • N-alkylation with chloromethyloxirane[4]
  • preparation of hydroindolepropynoate by chemo- and regioselective solvent-free ethynylation[1]
  • palladium- and copper-free cross-coupling of halopropynoates[5]
  • preparation of carbonylalkenyl indoles via coupling with dicarbonyl compounds[6]
  • 1:2 annelation of 4,5,6,7-tetrahydroindole with 1-benzoyl-2-phenylacetylene[7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrrole-2-dithiocarboxylates: Synthesis of 2-(1-Alkylthio-2-cyanoethenyl) pyrroles.
Sobenina LN, et al.
Tetrahedron, 51(14), 4223-4230 (1995)
Arcadi, A.; et al.
Advanced Synthesis & Catalysis, 348, 331-331 (2006)
Markova, M., V.; et al.
ARKIVOC (Gainesville, FL, United States), 57-57 (2008)
Sobenina, L. N.; et al.
Khim. Geterotsikl. Soedin., 39, 1113-1113 (2003)
Trofimov, B. A.; et al.
Tetrahedron Letters, 48, 4661-4661 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service