Skip to Content
Merck
All Photos(2)

Key Documents

324876

Sigma-Aldrich

N-(Triphenylphosphoranylidene)aniline

97%

Synonym(s):

Anilidenetriphenylphosphorane, Tetraphenylphosphine imide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$242.00

$242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$242.00

About This Item

Linear Formula:
(C6H5)3P=NC6H5
CAS Number:
Molecular Weight:
353.40
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$242.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

impurities

<0.5% denatured ethanol

solubility

chloroform: soluble 25 mg/mL, clear to slightly hazy (colorless to brown)

functional group

phosphine

SMILES string

c1ccc(cc1)N=P(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20NP/c1-5-13-21(14-6-1)25-26(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

PTLOPIHJOPWUNN-UHFFFAOYSA-N

Related Categories

General description

N-(Triphenylphosphoranylidene)aniline was reported as a new electrolyte additive for high performance lithium cobalt oxide (LiCoO2) electrodes during high voltage operations in lithium ion batteries.[1]

Application

The Staudinger Ligation: A High-Yield, Chemoselective, and Mild Synthetic Method

Intermediate for the preparation of isocyanates and isothiocyanates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-(triphenylphosphoranylidene) aniline as a novel electrolyte additive for high voltage LiCoO2 operations in lithium ion batteries.
Lee J-N, et al.
Electrochimica Acta, 56(14), 5195-5200 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service