Skip to Content
Merck
All Photos(3)

Key Documents

323276

Sigma-Aldrich

N-(Benzyloxycarbonyl)hydroxylamine

99%

Synonym(s):

N-Z-Hydroxylamine, Benzyl N-hydroxycarbamate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$64.40
25 G
$262.00

$64.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$64.40
25 G
$262.00

About This Item

Linear Formula:
C6H5CH2OCONHOH
CAS Number:
Molecular Weight:
167.16
Beilstein:
1074034
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$64.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

65-70 °C (lit.)

functional group

amine
phenyl

SMILES string

ONC(=O)OCc1ccccc1

InChI

1S/C8H9NO3/c10-8(9-11)12-6-7-4-2-1-3-5-7/h1-5,11H,6H2,(H,9,10)

InChI key

PQBSPTAPCMSZAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Danuta Gillner et al.
Bioorganic & medicinal chemistry letters, 19(22), 6350-6352 (2009-10-14)
The dapE-encoded N-succinyl-L,L-diaminopimelic acid desuccinylase (DapE) is a critical bacterial enzyme for the construction of the bacterial cell wall. A screen biased toward compounds containing zinc-binding groups (ZBG's) including thiols, carboxylic acids, boronic acids, phosphonates and hydroxamates has delivered a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service