Skip to Content
Merck
All Photos(2)

Documents

320625

Sigma-Aldrich

Ethyl 4-oxocyclohexanecarboxylate

97%

Synonym(s):

4-(Ethoxycarbonyl)cyclohexanone, Ethyl cyclohexanone-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O=)C6H9CO2C2H5
CAS Number:
Molecular Weight:
170.21
Beilstein:
2520501
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

150-152 °C/40 mmHg (lit.)

density

1.068 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1CCC(=O)CC1

InChI

1S/C9H14O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h7H,2-6H2,1H3

InChI key

ZXYAWONOWHSQRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the preparation of dopamine agonists and the skeleton of tetracyclic diterpenes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 35, 417-417 (1994)
Bioorganic & Medicinal Chemistry Letters, 3, 639-639 (1993)
Tetrahedron Letters, 35, 417-417 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service