Skip to Content
Merck
All Photos(4)

Key Documents

317039

Sigma-Aldrich

(+)-B-Methoxydiisopinocampheylborane

Synonym(s):

(+)-Diisopinocampheylmethoxyborane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$99.00
25 G
$307.00

$99.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$99.00
25 G
$307.00

About This Item

Empirical Formula (Hill Notation):
C21H37BO
CAS Number:
Molecular Weight:
316.33
Beilstein:
6370228
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$99.00


Check Cart for Availability

Request a Bulk Order

form

solid

SMILES string

COB([C@H]1CC2CC([C@@H]1C)C2(C)C)[C@H]3CC4CC([C@@H]3C)C4(C)C

InChI

1S/C21H37BO/c1-12-16-8-14(20(16,3)4)10-18(12)22(23-7)19-11-15-9-17(13(19)2)21(15,5)6/h12-19H,8-11H2,1-7H3/t12-,13-,14+,15+,16-,17-,18-,19-/m0/s1

InChI key

IAQXEQYLQNNXJC-JPDDNCELSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Both (+)- and (-)-forms are precursors to B-allyldiisopinocampheylborane derivatives, which are subsequently reacted with aldehydes to give homoallylic alcohols[1] and β-amino alcohols[2] of high enantiomeric excess.[3]
Reactant involved in:
  • Oxidative cyclization and alkylation for synthesis of ionomycin
  • Intramolecular conjugate addition for tetrahydropyran synthesis
  • Aldol addition and Suzuki coupling reactions
  • Allylation
  • Synthesis of alkanols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 56, 401-401 (1991)
Journal of the Chemical Society. Chemical Communications, 339-339 (1993)
Aldrichimica Acta, 20, 9-9 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service