Skip to Content
Merck
All Photos(1)

Key Documents

316768

Sigma-Aldrich

Methyllithium lithium iodide complex

1.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Li · LiI
Molecular Weight:
155.82
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

1.0 M in diethyl ether

bp

35 °C

density

0.909 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].[I-].[Li]C

InChI

1S/CH3.HI.2Li/h1H3;1H;;/q;;;+1/p-1

InChI key

LHJHNKBQWVOCIH-UHFFFAOYSA-M

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service