Skip to Content
Merck
All Photos(1)

Key Documents

31481

Sigma-Aldrich

cis-3,5-Diacetoxy-1-cyclopentene

≥98.0% (GC)

Synonym(s):

cis-4-Cyclopentene-1,3-diol diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O4
CAS Number:
Molecular Weight:
184.19
Beilstein:
1961566
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

solid

refractive index

n20/D 1.460

density

1.127 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)O[C@@H]1C[C@H](OC(C)=O)C=C1

InChI

1S/C9H12O4/c1-6(10)12-8-3-4-9(5-8)13-7(2)11/h3-4,8-9H,5H2,1-2H3/t8-,9+

InChI key

UKPIBFMHHUEUQR-DTORHVGOSA-N

General description

cis-3,5-Diacetoxy-1-cyclopentene is an organic building block.

Application

cis-3,5-Diacetoxy-1-cyclopentene may be employed as substrate to investigate the enantioselectivity of esterase EstA3 (obtained from a drinking water metagenome) and esterase EstCE1 (obtained from a soil metagenome).

Other Notes

Versatile building block; synthesis of carba-nucleosides ; Enzyme catalyzed hydrolysis to chiral acetoxy alcohol

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Elend et al.
Applied and environmental microbiology, 72(5), 3637-3645 (2006-05-05)
The metagenomes of uncultured microbial communities are rich sources for novel biocatalysts. In this study, esterase EstA3 was derived from a drinking water metagenome, and esterase EstCE1 was derived from a soil metagenome. Both esterases are approximately 380 amino acids
C.R. Johnson et al.
Tetrahedron Letters, 33, 7287-7287 (1992)
B.M. Trost et al.
Journal of the American Chemical Society, 114, 8745-8745 (1992)
D.R. Deardorff et al.
Tetrahedron Letters, 27, 1255-1255 (1986)
Senthil Kumar Rathnasamy et al.
Scientific reports, 10(1), 13356-13356 (2020-08-10)
The present study report for the first time on the one-pot production and purification of fibrinolytic protease from Bacillus cereus by extractive fermentation using natural deep eutectic solvents (NADES). Cheese whey was chosen as a sustainable low-cost production alternative yielding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service