Skip to Content
Merck
All Photos(3)

Documents

309303

Sigma-Aldrich

Magnesium perchlorate hexahydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2·6H2O
CAS Number:
Molecular Weight:
331.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

crystalline

reaction suitability

reagent type: oxidant

SMILES string

[Mg++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Mg.6H2O/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);;6*1H2/q;;+2;;;;;;/p-2

InChI key

MGBDNCBRZIMSDZ-UHFFFAOYSA-L

General description

Magnesium perchlorate hexahydrate is a highly hygroscopic metal hydrate that can be dehydrated to prepare anhydrous magnesium perchlorate. It can be used as an electrophilic catalyst for non-enzymic reduction of synthetic dihydropyridines. It may also be used in the formation of magnesium ion batteries.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water-Activated VOPO4 for Magnesium Ion Batteries
Ji X, et al.
Nano Letters, 18(10), 6441-6448 (2018)
Stability of perchlorate hydrates and their liquid solutions at the Phoenix landing site Mars
Chevrier VF, et al.
Geophysical Research Letters, 36(10) (2009)
Magnesium Perchlorate
Chakraborti AK, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 36(10) (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service