Skip to Content
Merck
All Photos(2)

Key Documents

308382

Sigma-Aldrich

Potassium hexafluorotitanate(IV)

Synonym(s):

Dipotassium hexafluorotitanate, Titanium potassium hexafluoride

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$97.80

$97.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$97.80

About This Item

Linear Formula:
K2(TiF6)
CAS Number:
Molecular Weight:
240.05
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$97.80


Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

reagent type: catalyst
core: titanium

SMILES string

[K+].[K+].F[Ti--](F)(F)(F)(F)F

InChI

1S/6FH.2K.Ti/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

RXCBCUJUGULOGC-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service