Skip to Content
Merck
All Photos(1)

Key Documents

308064

Sigma-Aldrich

2-(Trimethylsiloxy)furan

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$127.00
5 G
$560.00

$127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$127.00
5 G
$560.00

About This Item

Empirical Formula (Hill Notation):
C7H12O2Si
CAS Number:
Molecular Weight:
156.25
Beilstein:
1423066
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

34-35 °C/9 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C[Si](C)(C)Oc1ccco1

InChI

1S/C7H12O2Si/c1-10(2,3)9-7-5-4-6-8-7/h4-6H,1-3H3

InChI key

ILBCDLOQMRDXLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Silva López et al.
The Journal of organic chemistry, 70(9), 3654-3659 (2005-04-23)
[reaction: see text] A comprehensive scan of the transition state space for the reaction of 2-(trimethylsiloxy)furan and methacrolein (24 combinations) offered a satisfactory explanation of the high like diastereoselectivity obtained experimentally in the Mukaiyama vinylogous aldol reaction of these and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service