Skip to Content
Merck
All Photos(1)

Key Documents

307254

Sigma-Aldrich

Diisopropylphosphoramidous dichloride

95%

Synonym(s):

(Diisopropylamino)dichlorophosphine, Bis(isopropyl)aminodichlorophosphine, Dichloro(diisopropylamido)phosphorus, Dichloro(diisopropylamino)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NPCl2
CAS Number:
Molecular Weight:
202.06
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.485 (lit.)

density

1.096 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)N(C(C)C)P(Cl)Cl

InChI

1S/C6H14Cl2NP/c1-5(2)9(6(3)4)10(7)8/h5-6H,1-4H3

InChI key

UPPVRFOGRCBSJP-UHFFFAOYSA-N

Application

Reagent for the synthesis of peptides[1] and oligonucleotide analogues.[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 47, 2449-2449 (1991)
The Journal of Organic Chemistry, 59, 6687-6687 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service