Skip to Content
Merck
All Photos(3)

Documents

306215

Sigma-Aldrich

Poly(acrylic acid)

average Mv ~1,250,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

crosslinking

~0.1 % cross-linked

mol wt

average Mv ~1,250,000

viscosity

800-11,000 cP

transition temp

Tg 106 °C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence Glennon-Alty et al.
Acta biomaterialia, 9(4), 6041-6051 (2012-12-15)
Mesenchymal stem cells (MSCs) can generate chondrocytes in vitro, but typically need to be cultured as aggregates in the presence of transforming growth factor beta (TGF-β), which makes scale-up difficult. Here we investigated if polyacrylate substrates modelled on the functional
Imran Khimji et al.
Chemical communications (Cambridge, England), 49(13), 1306-1308 (2013-01-11)
The melting temperature of duplex DNA is much higher in polyanions than in non-ionic polymers with similar ionic strength, suggesting an additional electrostatic contribution on top of the excluded volume effect.
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)
Kang Zhong et al.
Carbohydrate polymers, 92(2), 1367-1376 (2013-02-13)
Phosphate rock (PHR), a traditional fertilizer, is abundant, but is hard to be utilized by plants. To improve the utilization of PHR, and to integrate water-retaining and controlled-release fertilizers, an agricultural superabsorbent polymer based on sulfonated corn starch/poly (acrylic acid)
Priscila Stona et al.
The journal of adhesive dentistry, 15(3), 221-227 (2013-04-06)
To examine the influence of 11.5% polyacrylic acid pretreatment on the interface and bond strength of self-adhesive resin cements (Maxcem Elite, RelyX Unicem, SeT) to dentin. Fifty-six third molars were randomly divided into seven groups: RelyX ARC as control (ARC)

Articles

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

We have discussed shortly the applications of 1:1 and 2:1 phyllosilicate material in membrane technology, as well as the basic geometric understanding of their layered structures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service