Skip to Content
Merck
All Photos(1)

Key Documents

305103

Sigma-Aldrich

Methyl 2-amino-5-chlorobenzoate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)CO2CH3
CAS Number:
Molecular Weight:
185.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

bp

168-170 °C/22 mmHg (lit.)

mp

66-68 °C (lit.)

functional group

chloro
ester

SMILES string

COC(=O)c1cc(Cl)ccc1N

InChI

1S/C8H8ClNO2/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4H,10H2,1H3

InChI key

IGHVUURTQGBABT-UHFFFAOYSA-N

General description

Methyl 2-amino-5-chlorobenzoate is a useful pharmaceutical intermediate. Crystal structure of methyl 2-amino-5-chlorobenzoate was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Bin Shi et al.
Acta crystallographica. Section E, Structure reports online, 66(Pt 11), o3025-o3025 (2010-01-01)
The title compound, C(8)H(8)ClNO(2), is almost planar, with an r.m.s. deviation of 0.0410 Å from the plane through the non-hydrogen atoms. In the crystal structure, inter-molecular N-H⋯O hydrogen bonds link the mol-ecules into chains along the b axis. An intra-molecular N-H⋯O

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service