Recommended Products
form
liquid
autoignition temp.
754 °F
concentration
1.0 M in THF
density
0.857 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
C\C(C)=C(\C)C
InChI
1S/C6H12/c1-5(2)6(3)4/h1-4H3
InChI key
WGLLSSPDPJPLOR-UHFFFAOYSA-N
General description
2,3-Dimethyl-2-butene undergoes ozonolysis in dark to yield hydroxyl radical. Reaction of ozone with 2,3-dimethyl 2-butene was investigated using a flow-tube interfaced to UV photoelectron spectrometer. It forms adduct with thianthrene cation radical tetrafluoroborate at 0°C and -15°C.
Application
Used for the preparation of thexylborane.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
1.4 °F
Flash Point(C)
-17 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of organic chemistry, 71(10), 3737-3742 (2006-05-06)
Thianthrene cation radical tetrafluoroborate (Th*+ BF4-) has been found to add to 2,3-dimethyl-2-butene (DMB) at 0 degrees C and -15 degrees C. The adduct, 2,3-dimethyl-2,3-(5,10-thianthreniumdiyl)butane ditetrafluoroborate (12), was isolated at -15 degrees C, and its 1H NMR spectrum was recorded
Environmental science & technology, 47(12), 6239-6246 (2013-05-15)
In order to investigate the heterogeneous oxidation kinetics of the herbicide terbuthylazine (TERB), a stable and reproducible generation system of "dark" hydroxyl radical in the gas phase was developed and optimized using a PTR-MS. TERB was adsorbed on silica particles
Photochemistry and photobiology, 61(6), 557-562 (1995-06-01)
The mechanism of photocycloaddition of 2'-deoxyuridine (1a) and thymidine (1b) to 2,3-dimethyl-2-butene (Bu) in acetonitrile by UV irradiation has been studied. The reciprocal quantum yield for the cycloaddition increased linearly with reciprocal concentrations of Bu in acetonitrile to give limiting
Biochemical and biophysical research communications, 117(2), 367-371 (1983-12-16)
Although indirect evidence has suggested that liver microsomal cytochrome P-450 can reductively dehalogenate several compounds to carbene metabolites, there has been no direct proof for the formation of these reactive species. We report in this paper that carbenes can be
Environmental science & technology, 41(7), 2357-2363 (2007-04-19)
We present a novel method for continuous, stable OH radical production for use in smog chamber studies, especially those focused on organic aerosol aging. Our source produces OH radicals from the reaction of 2,3-dimethyl-2-butene and ozone and is unique as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service