Skip to Content
Merck
All Photos(1)

Key Documents

303828

Sigma-Aldrich

Ethylmagnesium chloride solution

2.0 M in THF

Synonym(s):

Chloroethylmagnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$102.00
800 ML
$351.00

$102.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$102.00
800 ML
$351.00

About This Item

Linear Formula:
CH3CH2MgCl
CAS Number:
Molecular Weight:
88.82
Beilstein:
3647886
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$102.00


Available to ship on22 April 2025Details


Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in THF

density

0.978 g/mL at 25 °C

SMILES string

CC[Mg]Cl

InChI

1S/C2H5.ClH.Mg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1

InChI key

ZWNJJACKPNTXHY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ethylmagnesium chloride solution can be used:
  • In Cp2ZrCl2 catalyzed ethylcarboxylation of alkenes using carbon dioxide to corresponding carboxylic acids.[1]
  • In the preparation of cyclobutylmagnesium carbenoids by reacting with 1-chlorocyclobutyl p-tolyl sulfoxides.[2]

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclobutylmagnesium Carbenoids: Generation from 1-Chlorocyclobutyl p-Tolyl Sulfoxides with Ethylmagnesium Chloride, Properties, and Some Synthetic Uses
Satoh T, et al.
Synlett, 2010(02), 286-290 (2010)
Zirconocene-catalyzed sequential ethylcarboxylation of alkenes using ethylmagnesium chloride and carbon dioxide
Shao P, et al.
Chemical Communications (Cambridge, England), 51(30), 6640-6642 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service