Skip to Content
Merck
All Photos(1)

Key Documents

303127

Sigma-Aldrich

Triton X-405, reduced

reduced

Synonym(s):

Polyoxyethylene (40) isooctylcyclohexyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$714.00

$714.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$714.00

About This Item

Empirical Formula (Hill Notation):
C94H188O41
CAS Number:
Molecular Weight:
1974.47
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$714.00


Check Cart for Availability

Request a Bulk Order

Quality Level

mp

48-50 °C (lit.)

SMILES string

O(CCOCCOCCOCCO)CCOCCOCCOc1ccc(cc1)C(CC(C)(C)C)(C)C

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G E Tiller et al.
Analytical biochemistry, 141(1), 262-266 (1984-08-15)
The ultraviolet-light absorption and fluorescence of Triton X-100 were virtually eliminated by hydrogenation to its reduced cyclohexyl analog, RTX-100. The critical micelle concentration of RTX-100 was 12% higher than that of Triton X-100. RTX-100 and Triton X-100 were quite similar
J Fernandez et al.
Analytical biochemistry, 201(2), 255-264 (1992-03-01)
A procedure for the generation and isolation of internal peptide fragments for less than 10 micrograms of protein bound to either polyvinylidene difluoride (PVDF) or nitrocellulose membranes after electrophoretic transfer from sodium dodecyl sulfate-polyacrylamide gels (SDS-PAGE) is presented. This technique

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service