Skip to Content
Merck
All Photos(1)

Documents

302430

Sigma-Aldrich

Methylmagnesium bromide solution

1.0 M in dibutyl ether

Synonym(s):

Bromomethylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3MgBr
CAS Number:
Molecular Weight:
119.24
Beilstein:
3535220
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in dibutyl ether

density

0.856 g/mL at 25 °C

SMILES string

C[Mg]Br

InChI

1S/CH3.BrH.Mg/h1H3;1H;/q;;+1/p-1

InChI key

AVFUHBJCUUTGCD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Snahel Patel et al.
Journal of medicinal chemistry, 58(1), 401-418 (2014-10-24)
Dual leucine zipper kinase (DLK, MAP3K12) was recently identified as an essential regulator of neuronal degeneration in multiple contexts. Here we describe the generation of potent and selective DLK inhibitors starting from a high-throughput screening hit. Using proposed hinge-binding interactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service