Skip to Content
Merck
All Photos(1)

Key Documents

302147

Sigma-Aldrich

(S)-(+)-2-Methoxy-2-phenylethanol

98%

Synonym(s):

β-Methoxyphenethyl alcohol, (+)-β-Methoxyphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)CH2OH
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Assay

98%

form

liquid

optical activity

[α]19/D +133°, c = 1 in acetone

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.52 (lit.)

bp

65 °C/0.1 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

CO[C@H](CO)c1ccccc1

InChI

1S/C9H12O2/c1-11-9(7-10)8-5-3-2-4-6-8/h2-6,9-10H,7H2,1H3/t9-/m1/s1

InChI key

JDTUPLBMGDDPJS-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-2-Methoxy-2-phenylethanol can be used:
  • As a starting material for the synthesis of N-substituted 6,7-benzomorphan based opioid receptor agonists.[1]
  • As a ligand in the preparation of organolanthanoid complexes.[2]
  • As starting material for the synthesis of N-acetyl-O-((2S)-2-methoxy-2-phenylethyl)-L-serine, a serine proteases inhibitor.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing enzyme stereospecificity. Evaluation of β-alkoxy-α-amino acids with two stereocenters as inhibitors of serine proteases
Lee T and Jones JB
Tetrahedron, 51(27), 7331-7346 (1995)
Solid-state and solution structures of several new [LnCp′ 2 (OR)] 2 complexes (Ln= Pr, Yb) with chiral, oxygen-functionalized mu-alkoxide ligands
Steudel A, et al.
Journal of Organometallic Chemistry, 570(1), 89-96 (1998)
(2S)-N-2-methoxy-2-phenylethyl-6,7-benzomorphan compound (2S-LP2): Discovery of a biased mu/delta opioid receptor agonist
Pasquinucci L, et al.
European Journal of Medicinal Chemistry, 168, 189-198 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service