Skip to Content
Merck
All Photos(1)

Key Documents

300047

Sigma-Aldrich

2-Chloro-7-nitrofluorene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8ClNO2
CAS Number:
Molecular Weight:
245.66
MDL number:

Assay

95%

SMILES string

[O-][N+](=O)c1ccc-2c(Cc3cc(Cl)ccc-23)c1

InChI

1S/C13H8ClNO2/c14-10-1-3-12-8(6-10)5-9-7-11(15(16)17)2-4-13(9)12/h1-4,6-7H,5H2

InChI key

HJFWZNLUJDRROD-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service