Skip to Content
Merck
All Photos(1)

Key Documents

299146

Sigma-Aldrich

L-Methionine-(methyl-13C)

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$613.00
5 G
$2,260.00

$613.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$613.00
5 G
$2,260.00

About This Item

Linear Formula:
13CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
150.20
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$613.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

powder

optical activity

[α]20/D +23.1°, c = 1 in 1 M HCl

mp

284 °C (dec.) (lit.)

mass shift

M+1

storage temp.

room temp

SMILES string

[13CH3]SCCC(N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1+1

InChI key

FFEARJCKVFRZRR-YWQIHCTDSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

João M C Teixeira et al.
The FEBS journal, 286(6), 1230-1239 (2018-12-12)
Calcineurin is an essential calcium-activated serine/threonine phosphatase. The six NMR-observable methionine methyl groups in the catalytic domain of human calcineurin Aα (CNA) were assigned and used as reporters of the presence of potential cis-trans isomers in solution. Proline 84 is
Elizabeth I Parkinson et al.
Nature chemical biology, 15(11), 1049-1056 (2019-08-28)
Fosmidomycin and related molecules comprise a family of phosphonate natural products with potent antibacterial, antimalarial and herbicidal activities. To understand the biosynthesis of these compounds, we characterized the fosmidomycin producer, Streptomyces lavendulae, using biochemical and genetic approaches. We were unable
Fabia Canonica et al.
Science advances, 5(7), eaaw8478-eaaw8478 (2019-08-09)
The mechanisms underlying the biogenesis of the structurally unique, binuclear Cu1.5+•Cu1.5+ redox center (CuA) on subunit II (CoxB) of cytochrome oxidases have been a long-standing mystery. Here, we reconstituted the CoxB•CuA center in vitro from apo-CoxB and the holo-forms of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service