Skip to Content
Merck
All Photos(1)

Key Documents

297917

Sigma-Aldrich

Methyl 2,2-dichloro-2-methoxyacetate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$174.00

$174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$174.00

About This Item

Linear Formula:
Cl2C(OCH3)CO2CH3
CAS Number:
Molecular Weight:
172.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$174.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.45 (lit.)

bp

179-181 °C (lit.)

density

1.36 g/mL at 25 °C (lit.)

functional group

chloro
ester
ether

SMILES string

COC(=O)C(Cl)(Cl)OC

InChI

1S/C4H6Cl2O3/c1-8-3(7)4(5,6)9-2/h1-2H3

InChI key

FWIGIASHGJFYOL-UHFFFAOYSA-N

Application

Methyl 2,2-dichloro-2-methoxyacetate was used in the preparation of methyl (Z)-α-methoxyacrylates via stereospecific two-carbon homologation of aldehydes under Barbier conditions[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of methyl (Z)-a-methoxyacrylates via two-carbon homologation of aldehydes.
Baati R, et al.
Tetrahedron Letters, 50(4), 402-405 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service