Skip to Content
Merck
All Photos(1)

Key Documents

297836

Sigma-Aldrich

2-Chloro-4-fluorobenzoic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$163.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$163.00

About This Item

Linear Formula:
ClC6H3(F)CO2H
CAS Number:
Molecular Weight:
174.56
Beilstein:
1946215
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$163.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

181-183 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear to very slightly hazy, colorless to very faintly yellow

functional group

carboxylic acid
chloro
fluoro

SMILES string

OC(=O)c1ccc(F)cc1Cl

InChI

1S/C7H4ClFO2/c8-6-3-4(9)1-2-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

GRPWQLDSGNZEQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-4-fluorobenzoic acid was used in the preparation of 2-(2-chloro-4-fluorophenyl)-benzothiazole[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of novel benzothiazole derivatives against human cervical cancer cell lines.
Kini S, et al.
Indian Journal of Pharmaceutical Sciences, 69(1), 46-46 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service