Skip to Content
Merck
All Photos(1)

Key Documents

296880

Sigma-Aldrich

4-Methoxymandelic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH(OH)CO2H
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

108-111 °C (lit.)

SMILES string

COc1ccc(cc1)C(O)C(O)=O

InChI

1S/C9H10O4/c1-13-7-4-2-6(3-5-7)8(10)9(11)12/h2-5,8,10H,1H3,(H,11,12)

InChI key

ITECRQOOEQWFPE-UHFFFAOYSA-N

General description

Chiral separation of the enantiomers of 4-methoxymandelic acid has been reported by a new liquid chromatographic method[1]. The mechanism of veratryl alcohol-mediated oxidation of 4-methoxymandelic acid by lignin peroxidase has been studied by kinetic methods[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radical cation cofactors in lignin peroxidase catalysis.
P J Harvey et al.
Biochemical Society transactions, 23(2), 262-267 (1995-05-01)
M Tien et al.
The Journal of biological chemistry, 272(14), 8912-8917 (1997-04-04)
The mechanism of veratryl alcohol-mediated oxidation of 4-methoxymandelic acid by lignin peroxidase was studied by kinetic methods. For monomethoxylated substrates not directly oxidized by lignin peroxidase, veratryl alcohol has been proposed to act as a redox mediator. Our previous study
Jie Zhou et al.
Journal of Zhejiang University. Science. B, 14(7), 615-620 (2013-07-05)
A new liquid chromatographic method has been developed for the chiral separation of the enantiomers of mandelic acid and their derivatives 2-chloromandelic acid, 4-hydroxymandelic acid, 4-methoxymandelic acid, and 3,4,5-trismethoxymandelic acid. The enantiomers were separated by a CHIRALPAK(®) IC (250 mm×4.6
K Valli et al.
Biochemistry, 29(37), 8535-8539 (1990-09-18)
Lignin peroxidase (LiP), an extracellular heme enzyme from the lignin-degrading fungus Phanerochaete chrysosporium, catalyzes the H2O2-dependent oxidation of a variety of nonphenolic lignin model compounds. The oxidation of monomethoxylated lignin model compounds, such as anisyl alcohol (AA), and the role
P J Teunissen et al.
FEBS letters, 439(3), 219-223 (1998-12-09)
Poly R478, 4-methoxymandelic acid and oxalic acid were oxidized by lignin peroxidase (LiP) in the presence of the fungal metabolite 2-chloro-1,4-dimethoxybenzene (2Cl-14DMB), whereas no oxidation occurred in the absence of 2Cl-14DMB. These substrates clearly inhibited the consumption of 2Cl-14DMB and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service