Skip to Content
Merck
All Photos(1)

Key Documents

296368

Sigma-Aldrich

1,2-O-Isopropylidene-α-D-xylofuranose

99%

Synonym(s):

1,2-O-(1-Methylethylidene)-alpha-D-xylofuranose

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$376.00
100 G
$1,110.00

$376.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$376.00
100 G
$1,110.00

About This Item

Empirical Formula (Hill Notation):
C8H14O5
CAS Number:
Molecular Weight:
190.19
Beilstein:
81520
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

$376.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

optical activity

[α]25/D −19.2°, c = 1 in H2O

bp

112-114 °C/0.06 mmHg (lit.)

mp

69-71 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)O[C@H]2O[C@H](CO)[C@H](O)[C@H]2O1

InChI

1S/C8H14O5/c1-8(2)12-6-5(10)4(3-9)11-7(6)13-8/h4-7,9-10H,3H2,1-2H3/t4-,5+,6-,7-/m1/s1

InChI key

JAUQZVBVVJJRKM-XZBKPIIZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arie Gruzman et al.
Journal of medicinal chemistry, 51(24), 8096-8108 (2008-12-04)
Type 2 diabetes mellitus has reached epidemic proportions; therefore, the search for novel antihyperglycemic drugs is intense. We have discovered that D-xylose increases the rate of glucose transport in a non-insulin-dependent manner in rat and human myotubes in vitro. Due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service