Skip to Content
Merck
All Photos(1)

Key Documents

295353

Sigma-Aldrich

Hexafluoroacetone

97%

Synonym(s):

Hexafluoro-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CO
CAS Number:
Molecular Weight:
166.02
Beilstein:
607236
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Product 295353 is not currently sold in your country. Contact Technical Service

vapor density

1.65 (vs air)

vapor pressure

4525 mmHg ( 21.1 °C)

Assay

97%

bp

−26 °C (lit.)

mp

−129 °C (lit.)

SMILES string

FC(F)(F)C(=O)C(F)(F)F

InChI

1S/C3F6O/c4-2(5,6)1(10)3(7,8)9

InChI key

VBZWSGALLODQNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a 316 stainless steel diaphragm valve with a male 1/4" NPTF outlet thread (Z168637) installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Press. Gas Compr. Gas - Repr. 1B - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukihiro Kimura et al.
Photosynthesis research, 135(1-3), 23-31 (2017-05-12)
The light-harvesting 1 reaction center (LH1-RC) complex in the thermophilic purple sulfur bacterium Thermochromatium (Tch.) tepidum binds Ca ions as cofactors, and Ca-binding is largely involved in its characteristic Q
Fufeng Liu et al.
ACS chemical neuroscience, 10(1), 588-598 (2018-10-20)
The fibrillogenesis of amyloid-β protein (Aβ) is considered a crucial factor in the pathogenesis of Alzheimer's disease (AD). Hence, inhibiting Aβ fibrillogenesis is regarded as the primary therapeutic strategy for the prevention and treatment of AD. However, the development of
H Wille et al.
Journal of molecular biology, 259(4), 608-621 (1996-06-21)
The prion protein (PrP) undergoes a profound conformational change when the cellular isoform (PrPC) is converted into the scrapie form (PrPSc). Limited proteolysis of PrPsc produces PrP 27-30 which readily polymerizes into amyloid. To study the structure of PrP amyloid
V Prasanna et al.
Biochemistry, 37(19), 6883-6893 (1998-06-06)
Three synthetic peptides corresponding to distinct segments of the subunit interface of the dimeric enzyme thymidylate synthase (residues 17-38, N 22; residues 174-190, M 17; and residues 201-220, C 20) have been investigated for their ability to function as inhibitors
Jan Spengler et al.
The Journal of organic chemistry, 73(6), 2311-2314 (2008-02-22)
beta-Hydroxy acids were reacted with hexafluoroacetone and carbodiimides to give carboxy-activated six-membered lactones in good yields. On reaction with amines, the corresponding amides were obtained. We demonstrate the following applications of this protecting/activating strategy: preparation of carboxamides in solution and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service