Skip to Content
Merck
All Photos(1)

Documents

295310

Sigma-Aldrich

Chloroethane

≥99.7%

Synonym(s):

Ethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51
Beilstein:
1730751
EC Number:
MDL number:
UNSPSC Code:
12142100
eCl@ss:
39050105
PubChem Substance ID:

vapor density

2.22 (vs air)

vapor pressure

19.2 psi ( 20 °C)

Assay

≥99.7%

bp

12.3 °C (lit.)

mp

−139 °C (lit.)

density

0.921 g/mL at 0-4 °C (lit.)

SMILES string

CCCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - Repr. 1B

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-58.0 °F - closed cup

Flash Point(C)

-50.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Banerjee et al.
Journal of dentistry, 38(6), 475-479 (2010-03-13)
Compare clinical effectiveness of sodium bicarbonate and bioactive glass powders used for dental prophylaxis. 25 patients were allocated to either good or poor oral hygiene subgroups (n=50). Using a double-blind, split-mouth model, all patients underwent prophylaxis treatment on mandibular teeth;
David J Steward
Paediatric anaesthesia, 22(3), 275-277 (2011-10-26)
Charles Robson (Figure 1) was born in New Westminster, British Columbia (now a suburb of Vancouver), in 1884 and graduated in medicine from McGill University in Montreal in 1913. Having interned and with some anesthesia training at the Royal Victoria
Robert N Richards
Journal of cutaneous medicine and surgery, 13(5), 253-256 (2009-09-23)
Botulinum toxin injections are effective in the treatment of palmar and plantar hyperhidrosis, but discomfort has limited its use. To study the use of ethyl chloride medium-stream spray in reducing injection discomfort. We used ethyl chloride medium-stream spray, in conjunction
Lindsey L Cohen et al.
The Clinical journal of pain, 25(6), 490-494 (2009-06-23)
The purpose of this study was to evaluate the effectiveness of vapocoolant for preschoolers' immunization injection pain relief. Fifty-seven 4 to 6-year-old children were randomized into vapocoolant alone or typical care conditions. Pain was measured at the baseline and at
Y-G Im et al.
Journal of oral rehabilitation, 39(12), 931-940 (2012-09-22)
The aim of this study was to compare the effects of three cryotherapeutic modalities (ethyl chloride spraying, ice block rubbing and cold gel packing) on facial skin temperature. Thirty healthy volunteers (15 men, 15 women; mean age, 29·4 ± 3·2 years) participated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service