Skip to Content
Merck
All Photos(1)

Key Documents

293474

Sigma-Aldrich

4-Bromo-2-fluoroanisole

97%

Synonym(s):

4-Bromo-2-fluoro-1-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)OCH3
CAS Number:
Molecular Weight:
205.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.545 (lit.)

bp

84 °C/7 mmHg (lit.)

density

1.59 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

COc1ccc(Br)cc1F

InChI

1S/C7H6BrFO/c1-10-7-3-2-5(8)4-6(7)9/h2-4H,1H3

InChI key

DWNXGZBXFDNKOR-UHFFFAOYSA-N

General description

The IR and FT-IR spectra of 4-bromo-2-fluoroanisole have been investigated[1].

Application

4-Bromo-2-fluoroanisole has been used in the synthesis of:
  • 1,4-bis[(3′-fluoro-4′-n alkoxyphenyl)ethynyl]benzenes with n=1-12[2]
  • 7-fluoro-6-methoxy-1-methyl-2-naphthaldehyde[3]
  • 5,7-difluoro-6-methoxy-1-methyl-2-naphthaldehyde[3]
  • liquid crystals with terminal difluoromethoxy group and backbone of phenylbicyclohexane[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of molecular architectures for polymeric mesophase formation.
Pugh C, et al.
Macromolecular Symposia, 98(1) (1995)
Vibrational spectra, first hyperpolarizability, HOMO-LUMO analysis of 4-bromo 2-fluro anisole.
Arivazhagan M, et al.
Indian Journal of Pure and Applied Physics, 50(5), 299-307 (2012)
Rolf W Hartmann et al.
Journal of enzyme inhibition and medicinal chemistry, 19(2), 145-155 (2004-09-29)
Regioselectively fluorinated 1-(naphth-2-ylmethyl)imidazoles 1a-h have been synthesized starting from the corresponding (naphth-2-yl)methanols (2). 2a-d have been obtained by LiAlH4-promoted reduction of fluorinated 1-methyl-2-naphthaldehydes. The latter were easily prepared in fairly good overall yields by ceric ammonium nitrate (CAN)-promoted oxidative addition
Syntheses of Phenylbicyclohexane Liquid Crystals with Terminal Difluoromethoxy Group [J].
Li J, et al.
Fine Chemicals / ????, 12, 004-004 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service