Skip to Content
Merck
All Photos(2)

Key Documents

293458

Sigma-Aldrich

Pentachlorocyclopropane

technical grade, 90%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$18.70

$18.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$18.70

About This Item

Empirical Formula (Hill Notation):
C3HCl5
CAS Number:
Molecular Weight:
214.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$18.70


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

90%

refractive index

n20/D 1.5169 (lit.)

bp

55-56 °C/7 mmHg (lit.)

density

1.668 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClC1C(Cl)(Cl)C1(Cl)Cl

InChI

1S/C3HCl5/c4-1-2(5,6)3(1,7)8/h1H

InChI key

IACJMSLMMMSESC-UHFFFAOYSA-N

General description

Hydrogen-bonded complexes of pentachlorocyclopropane with the bases acetonitrile, ammonia, monomethylamine and dimethylamine have been isolated[1].

Application

Pentachlorocyclopropane has been used in the preparation of:
  • tetrachlorocyclopropene via reaction with 18M aqueous KOH at 80-85°C[2]
  • substituted phenyltrichlorocyclopropene derivatives[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Substituted Phenyltrichlorocyclopropene Derivatives Using Friedel-Crafts Reaction.
Choi S-K and Suk W-K.
Bull. Korean Chem. Soc., 2(3), 83-85 (1981)
Tetrachlorocyclopropene and hexachlorocyclopropane from pentachlorocyclopropane.
Tobey SW and West R.
Tetrahedron Letters, 4(18), 1179-1182 (1963)
Alexander B Baker et al.
The journal of physical chemistry. A, 109(37), 8280-8289 (2006-07-13)
Hydrogen-bonded complexes of pentachlorocyclopropane with the bases acetonitrile, ammonia, monomethylamine, and dimethylamine have been isolated and characterized for the first time in argon matrices at 16 K. Coordination of the proton of pentachlorocyclopropane (Pccp) to the electron donor (N) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service