Skip to Content
Merck
All Photos(1)

Documents

292478

Sigma-Aldrich

4,5-Difluoro-2-nitroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
174.10
Beilstein:
2723242
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

107-108 °C (lit.)

SMILES string

Nc1cc(F)c(F)cc1[N+]([O-])=O

InChI

1S/C6H4F2N2O2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H,9H2

InChI key

WDMCABATCGQAKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,5-Difluoro-2-nitroaniline has been used in the preparation of:
  • 2-chloro-5,6-difluorobenzimidazole
  • 1-(4,5-difluoro-2-nitrophenyl)pyrene via diazotization reaction with isoamyl nitrite in the presence of pyrene
  • 5-ethoxy-6-fluorobenzofuroxan

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Zou et al.
Journal of medicinal chemistry, 40(5), 811-818 (1997-02-28)
2-Chloro-5,6-difluorobenzimidazole (8) was prepared from 4,5-difluoro-2-nitroaniline (5) via successive reduction, cyclization, and diazotization reactions. 2-Chloro-5,6-dibromobenzimidazole (10) was obtained by a direct bromination of 2-chlorobenzimidazole (9) with bromine-water. 2-Chloro-5,6-diiodobenzimidazole (15) was synthesized by a stepwise transformation of the nitro functions of
Synthesis of fluorinated derivatives of benzo [k] fluoranthene and indeno [1, 2, 3-cd] pyrene and 8, 9-dihydro-8, 9-epoxybenzo [k] fluoranthene.
Rice JE, et al.
The Journal of Organic Chemistry, 53(8), 1775-1779 (1988)
Antonio Carta et al.
European journal of medicinal chemistry, 37(5), 355-366 (2002-05-15)
Thirty-six 6(7)-substituted-3-methyl- or 3-halogenomethyl-2-phenylthio-phenylsulphonyl-chloro-quinoxaline 1,4-dioxides belonging to series 3-6 were synthesised and submitted to a preliminary in vitro evaluation for antimycobacterial, anticandida and antibacterial activities. Antitubercular screening showed a generally good activity of 3-methyl-2-phenylthioquinoxaline 1,4-dioxides (3d,e,h-j) against Mycobacterium tuberculosis, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service