Skip to Content
Merck
All Photos(3)

Key Documents

291242

Sigma-Aldrich

Potassium oleate

40 wt. % paste in H2O

Synonym(s):

Oleic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOK
CAS Number:
Molecular Weight:
320.55
Beilstein:
4167152
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

concentration

40 wt. % paste in H2O

storage temp.

2-8°C

SMILES string

[K+].[H]\C(CCCCCCCC)=C(/[H])CCCCCCCC([O-])=O

InChI

1S/C18H34O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;

InChI key

MLICVSDCCDDWMD-KVVVOXFISA-M

Looking for similar products? Visit Product Comparison Guide

General description

Small-angle neutron-scattering study of microimulsions of potassium oleate, n-hexadecane, water and cosurfactant has been reported[1].

Application

Potassium oleate has been used in the preparation of:
  • nickel nanorods with a diameter of 8-10nm and a length of 100-200nm[2]
  • aqueous sols of calcium olcate[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation of nickel nanorods in water-in-oil microemulsion.
Ni X-M, et al.
Journal of Crystal Growth, 252(4), 612-617 (2003)
Small-angle neutron-scattering study of W/O n-hexadecane, potassium oleate, water and alcohol CnH2n+ 1OH (n= 5-8) microemulsions: effect of water concentration.
Caponetti E, et al.
Langmuir, 6(11), 1628-1634 (1990)
Precipitation phenomena of heavy metal soaps in aqueous solutions: I. Calcium oleate.
Matijevic E, et al.
Journal of Colloid and Interface Science, 22(5), 419-429 (1966)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service