Skip to Content
Merck
All Photos(1)

Key Documents

289760

Sigma-Aldrich

Pentaamminechlororuthenium(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ru(NH3)5Cl]Cl2
CAS Number:
Molecular Weight:
292.58
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
solubility:
H2O: slightly soluble(lit.)

grade

for analytical purposes

form

solid

reaction suitability

reagent type: catalyst
core: ruthenium

solubility

H2O: slightly soluble(lit.)

SMILES string

N.N.N.N.N.Cl[Ru](Cl)Cl

InChI

1S/3ClH.5H3N.Ru/h3*1H;5*1H3;/q;;;;;;;;+3/p-3

InChI key

HLAVJMYZYQTXAH-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Gibson et al.
Chemico-biological interactions, 53(3), 351-364 (1985-05-01)
A number of ruthenium complexes were tested for their ability to induce filamentation in Escherichia coli. These included monomeric and dimeric complexes with ruthenium in the II or III oxidation states, as well as mixed-valence complexes with ruthenium in the
J R Rubin et al.
Nucleic acids research, 11(18), 6571-6586 (1983-09-24)
A comparative study of the binding of square planar cis- and trans-[Pt(NH3)2Cl2] complexes and the octahedral [Ru(NH3)5(H2O)]3+ complex to tRNAphe from yeast was carried out by X-ray crystallography. Both of the carcinostatic compounds, cis-[Pt(NH3)2Cl2] and [Ru(NH3)5(H2O)]3+ show similarities in their
E John et al.
Acta crystallographica. Section C, Crystal structure communications, 48 ( Pt 9), 1574-1576 (1992-09-15)
[RuCl(NH3)5](HSO4)2.4H2O, M(r) = 487.88, triclinic, P1, a = 10.0422 (5), b = 14.1044 (7), c = 6.3273 (6) A, alpha = 100.369 (5), beta = 98.655 (6), gamma = 81.354 (4) degree, V = 864.5 (3) A3, Z = 2
T García et al.
Biosensors & bioelectronics, 27(1), 40-45 (2011-07-15)
A new electrochemical approach for an accurate quantification of DNA base pairs in genomic human DNA amplified by polymerase chain reaction (PCR) is described. The method is based on the immobilization of the sample (a thiolated DNA fragment) on the
J F Laliberté et al.
Journal of bioenergetics and biomembranes, 19(1), 69-81 (1987-02-01)
Ammineruthenium(III) complexes have been found to act as electron acceptors for the transplasmalemma electron transport system of animal cells. The active complexes hexaammineruthenium(III), pyridine pentammineruthenium(III), and chloropentaammineruthenium(III) range in redox potential (E'0) from 305 to -42 mV. These compounds also

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service