Skip to Content
Merck
All Photos(1)

Key Documents

289302

Sigma-Aldrich

Rubidium acetate

≥99.8% trace metals basis

Synonym(s):

Acetic acid rubidium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2Rb
CAS Number:
Molecular Weight:
146.53
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.8% trace metals basis

form

solid

SMILES string

[Rb+].CC([O-])=O

InChI

1S/C2H4O2.Rb/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

FOGKDYADEBOSPL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alana L Rister et al.
Journal of the American Society for Mass Spectrometry, 30(2), 248-255 (2018-11-11)
Steroids represent an interesting class of small biomolecules due to their use as biomarkers and their status as scheduled drugs. Although the analysis of steroids is complicated by the potential for many isomers, ion mobility spectrometry (IMS) has previously shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service