Skip to Content
Merck
All Photos(2)

Key Documents

289213

Sigma-Aldrich

Holmium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Holmium chloride hexahydrate, Holmium trichloride hexahydrate, Trichloroholmium hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$152.00
25 G
$376.00

$152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$152.00
25 G
$376.00

About This Item

Linear Formula:
HoCl3 · 6H2O
CAS Number:
Molecular Weight:
379.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

$152.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: holmium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

165 °C (dec.) (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ho](Cl)Cl

InChI

1S/3ClH.Ho.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

BUQFCXABQYNXLP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Ultrasound assisted one-pot synthesis of dihydropyrimidinones using holmium chloride as catalyst: This research highlights the use of holmium chloride hexahydrate as a catalyst for synthesizing dihydropyrimidinones, a class of compounds with significant pharmaceutical applications (Kakaei et al., 2015).

  • Easy synthesis and characterization of holmium-doped SPIONs: This study focuses on the material science aspect, where holmium-doped SPIONs are synthesized for potential use in various nanotechnological applications (Osial et al., 2018).

  • Synthesis and characterization of holmium-doped iron oxide nanoparticles: Describes the synthesis methods and potential applications of holmium-doped iron oxide nanoparticles, relevant for magnetic materials research (Bloemen et al., 2014).

  • Dy4, Dy5, and Ho2 Complexes of an N3O2 Aminophenol Donor: A Dy5-micro3-Peroxide Single Molecule Magnet: Examines the complexation of holmium in novel molecular architectures, important for understanding magnetic properties in materials chemistry (Corredoira-Vazquez et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service