Skip to Content
Merck
All Photos(3)

Key Documents

288039

Sigma-Aldrich

Methyltin trichloride

97%

Synonym(s):

Trichloromethylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SnCl3
CAS Number:
Molecular Weight:
240.10
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

171 °C (lit.)

mp

48-51 °C (lit.)

SMILES string

C[Sn](Cl)(Cl)Cl

InChI

1S/CH3.3ClH.Sn/h1H3;3*1H;/q;;;;+3/p-3

InChI key

YFRLQYJXUZRYDN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

thymus,Nervous system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lina Huang et al.
International journal of analytical chemistry, 2020, 1975471-1975471 (2020-04-16)
Organic tin compounds (OTCs), a group of high-risk hazardous substances, are highly concerned in safety regulation of consumer products especially for toys because they can cause serious damage to organs after prolonged or repeated exposure. Gastric juice migration is nowadays
Hiroshi Urushitani et al.
Aquatic toxicology (Amsterdam, Netherlands), 199, 103-115 (2018-04-06)
Two cDNAs of RXR were isolated, for the first time, from the ivory shell, Babylonia japonica, and the transcriptional activities were tested in vitro to compare with other gastropod (Thais clavigera and Nucella lapillus) RXR isoforms. The transcriptional activities of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service