Skip to Content
Merck
All Photos(1)

Key Documents

286486

Sigma-Aldrich

Dodecanoic anhydride

≥98%

Synonym(s):

Lauric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10CO]2O
CAS Number:
Molecular Weight:
382.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98%

form

crystals

mp

41-43 °C (lit.)

SMILES string

CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC

InChI

1S/C24H46O3/c1-3-5-7-9-11-13-15-17-19-21-23(25)27-24(26)22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3

InChI key

NWADXBLMWHFGGU-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The immunogenecity of the material made by coupling dodecanoic anhydride to NH2 groups was studied[1].

Application

Dodecanoic anhydride was used to chemically conjugate tumor cell membrane in carbonate buffer[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Šedová et al.
Carbohydrate polymers, 216, 63-71 (2019-05-03)
The effect of hydrazide linkers on the formation and mechanical properties of hyaluronan hydrogels was intensively evaluated. The reaction kinetics of hydrazone formation was monitored by NMR spectroscopy under physiological conditions where polyaldehyde hyaluronan (unsaturated: ΔHA-CHO, saturated: HA-CHO) was reacted
R L Hunter et al.
Journal of the National Cancer Institute, 54(5), 1157-1163 (1975-05-01)
The covalent conjugation of fatty acid to a tumor cell membrane preparation transformed it from an antigen that enhanced tumor growth to one that suppressed it. A crude cell membrane preparation was made by sequential hypertonic and hypotonic salt extraction
Gordon D Sproul
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 49(4), 241-254 (2019-12-29)
Amino acids and peptides have been demonstrated to form lipoamino acids and lipopeptides under presumed prebiotic conditions, and readily form liposomes. Of the common nucleobases, adenine forms a liponucleobase even below 100 °C. Adenine as well as other nucleobases can also
S B Singh et al.
Journal of immunology (Baltimore, Md. : 1950), 120(3), 734-738 (1978-03-01)
A lipid conjugate of BSA was made by coupling dodecylamine to the COOH groups of BSA (DA-BSA). Studies on the immunogenicity of this material and one made by coupling dodecanoic anhydride to NH2 groups (D-BSA) demonstrated that cell-mediated immunity could
C Bastiancich et al.
Journal of controlled release : official journal of the Controlled Release Society, 225, 283-293 (2016-02-02)
The local delivery of chemotherapeutic agents is a very promising strategy for the treatment of glioblastoma (GBM). Gemcitabine is a chemotherapeutic agent that has a different mechanism of action compared to alkylating agents and shows excellent radio-sensitizing properties. So, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service