Skip to Content
Merck
All Photos(1)

Key Documents

285587

Sigma-Aldrich

Diethyl fluoromalonate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$137.00

$137.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$137.00

About This Item

Linear Formula:
FCH(COOC2H5)2
CAS Number:
Molecular Weight:
178.16
Beilstein:
1775686
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$137.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

121-122 °C/30 mmHg (lit.)

density

1.129 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)C(F)C(=O)OCC

InChI

1S/C7H11FO4/c1-3-11-6(9)5(8)7(10)12-4-2/h5H,3-4H2,1-2H3

InChI key

GOWQBFVDZPZZFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of diethyl fluoromalonate with electron-poor and electron-rich, sterically hindered and unhindered aryl bromides and chlorides were studied[1].

Application

The kinetic parameters of diethyl fluoromalonate in hemolysates were used to study carboxylesterase activities in complex systems[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L Mendz et al.
Archives of biochemistry and biophysics, 305(2), 252-260 (1993-09-01)
Eight fluorinated compounds were tested as putative probes to measure carboxylesterase activity employing 19F nuclear magnetic resonance spectroscopy. The method takes advantage of the sensitivity of fluorine resonances to the changes in the chemical bonding in the covalent structures where
Neil A Beare et al.
The Journal of organic chemistry, 67(2), 541-555 (2002-01-19)
Palladium-catalyzed reactions of aryl bromides and chlorides with two common stabilized carbanions-enolates of dialkyl malonates and alkyl cyanoesters-are reported. An exploration of the scope of these reactions was conducted, and the processes were shown to occur in a general fashion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service