Skip to Content
Merck
All Photos(1)

Documents

284734

Sigma-Aldrich

Indole-3-carboxylic acid

ReagentPlus®, 99%

Synonym(s):

β-Indolylcarboxylic acid, 3-Carboxyindole, 3-Indole formic acid, 3-Indolylcarboxylic acid, Indole-β-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Beilstein:
129435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

solid

mp

232-234 °C (dec.) (lit.)

solubility

95% ethanol: soluble 5%, clear to slightly hazy, light yellow to yellow

SMILES string

OC(=O)c1c[nH]c2ccccc12

InChI

1S/C9H7NO2/c11-9(12)7-5-10-8-4-2-1-3-6(7)8/h1-5,10H,(H,11,12)

InChI key

KMAKOBLIOCQGJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The structures of the derivatives of indole-3-carboxylic acid were studied using gas chromatography with high resolution mass spectrometry (GC-HRMS), ultra-high performance liquid chromatography in combination with high resolution tandem mass spectrometry (UHPLC-HRMS), nuclear magnetic resonance spectroscopy (NMR) and Fourier transform infrared spectroscopy (FT-IR).

Application

Reactant for preparation of:
  • Anticancer agents
  • Derivatives of amino acids and peptides
  • Serotonin 5-HT4 receptor antagonists
  • Primary acylureas
  • Inhibitors of Gli1-mediated transcription in the Hedgehog pathway
  • Serotonin 5-HT6 antagonists
  • Very Late Antigen-4 (VLA-4) sntagonists
  • EphB3 receptor tyrosine kinase inhibitors
  • Potential Therapeutic Agent for Alzheimer′s Disease
  • Vinyl ester pseudotripeptide proteasome inhibitors

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vadim Shevyrin et al.
Forensic science international, 232(1-3), 1-10 (2013-09-24)
By means of gas chromatography with high resolution mass spectrometry (GC-HRMS), ultra-high performance liquid chromatography in combination with high resolution tandem mass spectrometry (UHPLC-HRMS), nuclear magnetic resonance spectroscopy (NMR) and Fourier transform infrared spectroscopy (FT-IR), structure of a series from
Xuejin Zhao et al.
Viruses, 13(8) (2021-08-29)
Influenza A viruses are serious zoonotic pathogens that continuously cause pandemics in several animal hosts, including birds, pigs, and humans. Indole derivatives containing an indole core framework have been extensively studied and developed to prevent and/or treat viral infection. This
Mu-Yang Wang et al.
Journal of integrative plant biology, 54(7), 471-485 (2012-05-26)
Camalexin (3-thiazol-2'-yl-indole) is the major phytoalexin found in Arabidopsis thaliana. Several key intermediates and corresponding enzymes have been identified in camalexin biosynthesis through mutant screening and biochemical experiments. Camalexin is formed when indole-3-acetonitrile (IAN) is catalyzed by the cytochrome P450
M U Ahmad et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 23(9), 841-847 (1985-09-01)
The nitrosation of gramine, a tertiary amine alkaloid present in barley malt, was carried out by reaction with sodium nitrite in buffered acetic acid (pH 3.4) for 1 hr at room temperature. Two major non-volatile products of the nitrosation reaction
Jamil A Shilpi et al.
Fitoterapia, 81(6), 536-539 (2010-01-26)
Phytochemical investigation of the stem bark of Trewia nudiflora led to the isolation of two new cardenolides, trewianin (1) and trewioside (2), along with scopoletin (3) and indole-3-carboxylic acid (4). Structures were established by 1D- and 2D NMR spectroscopy, HR-MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service