Skip to Content
Merck
All Photos(1)

Key Documents

284718

Sigma-Aldrich

(−)-Cotinine

98%

Synonym(s):

(S)-1-Methyl-5-(3-pyridyl)-2-pyrrolidinone, S(−)-1-Methyl-5-(3-pyridyl)-2-pyrrolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O
CAS Number:
Molecular Weight:
176.22
Beilstein:
83099
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −21°, c = 1 in ethanol

bp

250 °C/150 mmHg (lit.)

mp

40-42 °C (lit.)

SMILES string

CN1[C@@H](CCC1=O)c2cccnc2

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1

InChI key

UIKROCXWUNQSPJ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Major metabolite of nicotine in humans.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Florescu et al.
Therapeutic drug monitoring, 29(4), 437-446 (2007-08-02)
Exposure to environmental tobacco smoke (ETS) is most often estimated using questionnaires, but they are unreliable. Biomarkers can provide valid information on ETS exposure, the preferred biomarker being cotinine. However, no reference range of hair cotinine exists to distinguish among
Satu Kalliola et al.
The Journal of allergy and clinical immunology, 131(3), 730-735 (2013-02-06)
Exposure to tobacco smoke is a well-known risk factor for childhood asthma and reduced lung function, but the effect on airway inflammation in preschool-aged children is unclear. To examine the effect of parental smoking on lung function and fractional concentration
Esther García-Esquinas et al.
Pediatrics, 131(5), e1415-e1423 (2013-04-10)
Active smoking and secondhand smoke (SHS) are known risk factors for kidney disease in adults. We evaluated the association between exposure to active smoking or SHS and kidney function in US adolescents. This is a cross-sectional study in 7516 adolescents
Sharon Saydah et al.
Pediatrics, 131(3), e679-e686 (2013-02-20)
To determine the risk of mortality associated with cardiometabolic risk factors in a national sample of adolescents and young adults. Prospective study of participants in the third NHANES (1988-1994), aged 12 to 39 years at the time of the survey
R Machaalani et al.
Toxicology and applied pharmacology, 276(3), 204-212 (2014-03-13)
Smoking during pregnancy is associated with low birth weight, premature delivery, and neonatal morbidity and mortality. Nicotine, a major pathogenic compound of cigarette smoke, binds to the nicotinic acetylcholine receptors (nAChRs). A total of 16 nAChR subunits have been identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service