Skip to Content
Merck
All Photos(1)

Key Documents

284521

Sigma-Aldrich

Ethyl phenyl sulfide

97%

Synonym(s):

Ethylthiobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SC2H5
CAS Number:
Molecular Weight:
138.23
Beilstein:
1905731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.566 (lit.)

bp

204-205 °C (lit.)

density

1.021 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCSc1ccccc1

InChI

1S/C8H10S/c1-2-9-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

AEHWKBXBXYNPCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The activation parameters involved in the oxidation of ethyl phenyl sulfide in tert-butyl alcohol were studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Bennett et al.
Inorganic chemistry, 40(13), 2996-3001 (2001-06-12)
A detailed mechanism for the oxidation of aryl sulfides by peroxymonocarbonate ion in cosolvent/water media is described. Kinetic studies were performed to characterize the transition state, including a Hammett correlation and variation of solvent composition. The results are consistent with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service