Skip to Content
Merck
All Photos(2)

Key Documents

284491

Sigma-Aldrich

(R)-(+)-2-Amino-3-phenyl-1-propanol

98%

Synonym(s):

D-Phenylalaninol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CH2OH
CAS Number:
Molecular Weight:
151.21
Beilstein:
2208239
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]/D +22.8°, c = 1.2 in 1 M HCl

mp

93-95 °C (lit.)

functional group

amine
hydroxyl
phenyl

SMILES string

N[C@@H](CO)Cc1ccccc1

InChI

1S/C9H13NO/c10-9(7-11)6-8-4-2-1-3-5-8/h1-5,9,11H,6-7,10H2/t9-/m1/s1

InChI key

STVVMTBJNDTZBF-SECBINFHSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

W W Mak et al.
Canadian journal of biochemistry, 58(12), 1421-1429 (1980-12-01)
The activities of a range of phenylalaninol-related compounds on capping of concanavalin A and induction of rounding of Chinese hamster ovary tsHl cells, as well as on the fluidity of phosphatidylcholine-cholesterol (1:1) liposomes, have been examined. These compounds include phenylalaninol
Gretta C M'bitsi-Ibouily et al.
Scientific reports, 9(1), 4146-4146 (2019-03-13)
Sulpiride (SPR) is a selective antagonist of central dopamine receptors but has limited clinical use due to its poor pharmacokinetics. The aim of this study was to investigate how metal ligation to SPR may improve its solubility, intestinal permeability and
M O Blondel et al.
Biochemical and biophysical research communications, 150(3), 979-986 (1988-02-15)
The illumination of Escherichia coli cells with UVA light, 320 nm less than or equal to lambda less than or equal to 380 nm, triggers a transient growth and division delay. The built-in 4-thiouridine chromophore which absorbs light at 340
W W Mak et al.
Canadian journal of biochemistry, 58(9), 737-744 (1980-09-01)
This study describes a rounding reaction induced in mammalian cells by the addition of phenylalaninol. In the Chinese hamster ovary tsH1 line the rounding occurred rapidly with a half time of 1 min at 25 mM phenylalaninol. After the removal
Violetta Constantinou-Kokotou et al.
Journal of peptide science : an official publication of the European Peptide Society, 11(7), 431-435 (2005-01-07)
2-Oxoamides based on long chain beta-amino acids were synthesized. 1-Benzyl substituted long chain amines, needed for such synthesis, were synthesized starting from Boc-phenylalaninol. The oxidative conversion of a phenyl group to a carboxyl group was used as the key transformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service