Skip to Content
Merck
All Photos(1)

Key Documents

283711

Sigma-Aldrich

N,N,2,4,6-Pentamethylaniline

98%

Synonym(s):

N,N-Dimethylmesidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2N(CH3)2
CAS Number:
Molecular Weight:
163.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.512 (lit.)

bp

213-215 °C (lit.)

density

0.907 g/mL at 25 °C (lit.)

SMILES string

CN(C)c1c(C)cc(C)cc1C

InChI

1S/C11H17N/c1-8-6-9(2)11(12(4)5)10(3)7-8/h6-7H,1-5H3

InChI key

JZBZLRKFJWQZHU-UHFFFAOYSA-N

General description

N,N,2,4,6-Pentamethylaniline participates in condensation of dimeric 3,4,6-tri-O-acetyl-2-deoxy-2-nitroso-α-D-glucopyranosyl chloride with benzyl alcohol to yield an oximino glycoside.

Application

N,N,2,4,6-Pentamethylaniline was used in the synthesis of an oximino glycoside that was reduced with lithium aluminum hydride to benzyl 2-amino-2-deoxy-α-D-gluco- and -mannopyranoside hydrochloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a β-D-linked disaccharide from dimeric tri-O-acetyl-2-deoxy-2-nitroso-α-D-glucopyranosyl chloride.
Miyai K and Jeanloz RW.
Carbohydrate Research, 21(1), 45-55 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service