Skip to Content
Merck
All Photos(1)

Key Documents

283029

Sigma-Aldrich

2,6-Bis(4-azidobenzylidene)-4-methylcyclohexanone

contains 20-45% water as stabilizer, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N6O
CAS Number:
Molecular Weight:
370.41
Beilstein:
3017027
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

contains

20-45% water as stabilizer

reaction suitability

reaction type: click chemistry

mp

124-126 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1CC(=C/c2ccc(cc2)N=[N+]=[N-])\C(=O)\C(C1)=C\c3ccc(cc3)N=[N+]=[N-]

InChI

1S/C21H18N6O/c1-14-10-17(12-15-2-6-19(7-3-15)24-26-22)21(28)18(11-14)13-16-4-8-20(9-5-16)25-27-23/h2-9,12-14H,10-11H2,1H3/b17-12+,18-13+

InChI key

MLIWQXBKMZNZNF-PWDIZTEBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Bis(4-azidobenzylidene)-4-methylcyclohexanone has been used:
  • in the preparation of homogeneous pseudo-interpenetrating polymer networks[1]
  • as photosensitive agent in the preparation of photosensitive benzocyclobutene-functionalized siloxane thermosets[2]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of photosensitive benzocyclobutene?functionalized siloxane thermosets.
Zuo X, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47(22), 6246-6258 (2009)
Tuning the free volume cavities of polyimide membranes via the construction of pseudo-interpenetrating networks for enhanced gas separation performance.
Low BT, et al.
Macromolecules, 42(18), 7042-7054 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service