Skip to Content
Merck
All Photos(1)

Key Documents

282448

Sigma-Aldrich

Propionic acid-1-13C

99 atom % 13C

Synonym(s):

Propanoic acid-1-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$500.00

$500.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$500.00

About This Item

Linear Formula:
CH3CH213CO2H
CAS Number:
Molecular Weight:
75.07
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$500.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

technique(s)

mass spectrometry (MS): suitable

bp

141 °C (lit.)

mp

-24--23 °C (lit.)

density

0.993 g/mL at 25 °C

mass shift

M+1

SMILES string

CC[13C](O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i3+1

InChI key

XBDQKXXYIPTUBI-LBPDFUHNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Huclier-Markai et al.
Rapid communications in mass spectrometry : RCM, 32(16), 1331-1343 (2018-05-29)
Natural organic matter (NOM) is present in the environment and could influence the migration of heavy metals/radionuclides. The dissolved fraction of NOM (DOM) is usually quantified using total organic carbon analysis or UV-visible spectrometry. Nonetheless, analysis using pattern recognition cannot

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service