Skip to Content
Merck
All Photos(1)

Key Documents

280984

Sigma-Aldrich

1-Methylxanthine

98%

Synonym(s):

2,6-Dihydroxy-1-methylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein:
171507
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

mp

≥300 °C

SMILES string

CN1C(=O)Nc2nc[nH]c2C1=O

InChI

1S/C6H6N4O2/c1-10-5(11)3-4(8-2-7-3)9-6(10)12/h2H,1H3,(H,7,8)(H,9,12)

InChI key

MVOYJPOZRLFTCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Caffeine metabolite.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong-Yun Jeong et al.
International journal of pharmaceutics, 372(1-2), 132-139 (2009-01-27)
Most of methylxanthine derivatives including caffeine have been known to radiosensitize cancer cells, but the obstacles such as toxicity, request of high dose and poor solubility hinder their preclinical evaluations and clinical applications. In this study, we evaluated the efficacy
Catherine M Wheatley et al.
American journal of physiology. Endocrinology and metabolism, 287(4), E804-E809 (2004-06-24)
Exercise and insulin increase muscle glucose uptake by different mechanisms and also increase capillary recruitment, which is proposed to facilitate access for hormones and nutrients. The genetically obese Zucker rat shows impaired insulin- but not contraction-mediated glucose uptake in muscle.
Stephen Rattigan et al.
Exercise and sport sciences reviews, 33(1), 43-48 (2005-01-11)
Blood flow to contracting muscles is increased rapidly and is redistributed within the muscle. Recent research has highlighted that insulin causes similar changes, but these are absent in insulin resistance, resulting in impaired insulin-mediated muscle glucose uptake. Understanding the mechanisms
Hyewon Youn et al.
International journal of radiation biology, 85(2), 167-174 (2009-03-13)
To determine the efficacy of a caffeine derivative 1-methylxanthine (1-MTX) in increasing radiosensitivity of cancer cells and elucidate the underlying mechanisms in vitro. RKO human colorectal cancer cells carrying wild type protein 53 kDa (p53) were incubated with 3 mM
P St-Pierre et al.
Diabetes, obesity & metabolism, 14(8), 753-761 (2012-03-21)
Exercise and insulin each increase microvascular blood flow and enhance glucose disposal in skeletal muscle. We have reported that insulin-mediated microvascular recruitment in a diet-induced model of insulin resistance (high-fat feeding for 4 weeks) is markedly impaired; however, the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service